accessibility menu, dialog, popup

UserName

CAS RN 59443-15-1

IUPAC Name: 1-(3-bromo-5-chloro-2-hydroxyphenyl)ethan-1-one
Synonyms: 1-(3-bromo-5-chloro-2-hydroxyphenyl)ethanone
Molecular Weight (g/mol): 249.49
Molecular Formula: C8H6BrClO2
InChi Key: FFAVKFQPEOGJOA-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(Cl)=CC(Br)=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results