missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7123749
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47861G 1 g
B47865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47861G Supplier TCI America Supplier No. B47861G
Only null left

Chemical Identifiers

CAS 101420-81-9
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.02
MDL Number MFCD04117949
InChI Key KKPPNEJUUOQRLE-UHFFFAOYSA-N
PubChem CID 16218087
IUPAC Name 3-bromo-4-nitrobenzoic acid
SMILES OC(=O)C1=CC(Br)=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 203°C
Quantity 1 g
Synonym 3-bromo-4-nitrobenzoic acid, 3-bromo-4-nitrobenzoicacid, benzoic acid,3-bromo-4-nitro, benzoic acid, 3-bromo-4-nitro, 3-bromo-4-nitrobenzoic acid 97, pubchem16301, acmc-209x3e, 4-nitro-3-bromobenzoic acid, 3-bromo-4-nitro-benzoic acid
Formula Weight 246.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-nitrobenzoic Acid