missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-nitrobenzoic acid, 97%

Catalog No. AAH2726803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2726803 1 g
AAH2726806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2726803 Supplier Thermo Scientific Chemicals Supplier No. H2726803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 101420-81-9 | C7H4BrNO4 | 246.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 101420-81-9
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.02
MDL Number MFCD04117949
InChI Key KKPPNEJUUOQRLE-UHFFFAOYSA-N
Synonym 3-bromo-4-nitrobenzoic acid, 3-bromo-4-nitrobenzoicacid, benzoic acid,3-bromo-4-nitro, benzoic acid, 3-bromo-4-nitro, 3-bromo-4-nitrobenzoic acid 97, pubchem16301, acmc-209x3e, 4-nitro-3-bromobenzoic acid, 3-bromo-4-nitro-benzoic acid
PubChem CID 16218087
IUPAC Name 3-bromo-4-nitrobenzoic acid
SMILES OC(=O)C1=CC(Br)=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 200°C to 204°C
Quantity 1 g
Formula Weight 246.02
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only