missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4-methylthiophene 93.0+%, TCI America™
Supplier: TCI America B154125G
Specifications
| 3-Bromo-4-methylthiophene | |
| Yellow | |
| C5H5BrS | |
| 25 g | |
| MBUSOPVRLCFJCS-UHFFFAOYSA-N | |
| 3-bromo-4-methylthiophene | |
| 2734935 | |
| ≥93.0% (GC) |
| 30318-99-1 | |
| 180°C | |
| MFCD00130084 | |
| 3-bromo-4-methyl-thiophene, thiophene, 3-bromo-4-methyl, pubchem21832, acmc-209her, 3-bromo-4-methylthiophen, 4-bromo-3-methylthiophene, bidd:gt0486, 3-bromo-4-methylthiophene | |
| CC1=CSC=C1Br | |
| 177.059 | |
| 177.06 | |
| Liquid |
Chemical Identifiers
| 30318-99-1 | |
| 177.059 | |
| MBUSOPVRLCFJCS-UHFFFAOYSA-N | |
| 2734935 | |
| CC1=CSC=C1Br |
| C5H5BrS | |
| MFCD00130084 | |
| 3-bromo-4-methyl-thiophene, thiophene, 3-bromo-4-methyl, pubchem21832, acmc-209her, 3-bromo-4-methylthiophen, 4-bromo-3-methylthiophene, bidd:gt0486, 3-bromo-4-methylthiophene | |
| 3-bromo-4-methylthiophene |
Safety and Handling
TSCA : No