Learn More
3-Bromo-4-methylthiophene, 95%, Thermo Scientific Chemicals
$238.57 - $238.57
Chemical Identifiers
| CAS | 30318-99-1 |
|---|---|
| Molecular Formula | C5H5BrS |
| Molecular Weight (g/mol) | 177.06 |
| MDL Number | MFCD00130084 |
| InChI Key | MBUSOPVRLCFJCS-UHFFFAOYSA-N |
| Synonym | 3-bromo-4-methyl-thiophene, thiophene, 3-bromo-4-methyl, pubchem21832, acmc-209her, 3-bromo-4-methylthiophen, 4-bromo-3-methylthiophene, bidd:gt0486, 3-bromo-4-methylthiophene |
| PubChem CID | 2734935 |
| IUPAC Name | 3-bromo-4-methylthiophene |
| SMILES | CC1=CSC=C1Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC379960050
|
Thermo Scientific Chemicals
379960050 |
5 g | Glass bottle |
Each for $238.57
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 30318-99-1 | |
| 177.06 | |
| MBUSOPVRLCFJCS-UHFFFAOYSA-N | |
| 2734935 | |
| CC1=CSC=C1Br |
| C5H5BrS | |
| MFCD00130084 | |
| 3-bromo-4-methyl-thiophene, thiophene, 3-bromo-4-methyl, pubchem21832, acmc-209her, 3-bromo-4-methylthiophen, 4-bromo-3-methylthiophene, bidd:gt0486, 3-bromo-4-methylthiophene | |
| 3-bromo-4-methylthiophene |
Specifications
| 30318-99-1 | |
| 100.0 | |
| 1.5840g/mL | |
| 78°C | |
| 94% min. (GC) | |
| C5H5BrS | |
| MFCD00130084 | |
| 1.584 | |
| MBUSOPVRLCFJCS-UHFFFAOYSA-N | |
| 3-bromo-4-methylthiophene | |
| 2734935 | |
| 95% | |
| 3-Bromo-4-methylthiophene |
| 94.0 | |
| Colorless to Yellow | |
| 179°C to 183°C (770.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.5770 to 1.579 | |
| 5 g | |
| 3-bromo-4-methyl-thiophene, thiophene, 3-bromo-4-methyl, pubchem21832, acmc-209her, 3-bromo-4-methylthiophen, 4-bromo-3-methylthiophene, bidd:gt0486, 3-bromo-4-methylthiophene | |
| CC1=CSC=C1Br | |
| 177.06 | |
| 177.06 | |
| Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Call a POISON CENTER or doctor/physician if
GHS Signal Word: Warning
RUO – Research Use Only