missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methoxybenzoic acid, 98+%

Catalog No. p-7020630
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1265306 5 g
AAA1265314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1265306 Supplier Thermo Scientific Chemicals Supplier No. A1265306
Only null left

CAS: 99-58-1 | C8H6BrO3 | 230.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-58-1
Molecular Formula C8H6BrO3
Molecular Weight (g/mol) 230.04
MDL Number MFCD00020295
InChI Key BBPZABXVRBFWGD-UHFFFAOYSA-M
Synonym 3-bromo-p-anisic acid, benzoic acid, 3-bromo-4-methoxy, 3-bromo-4-methoxybenzoicacid, akos bbb/210, rarechem al bo 0751, timtec-bb sbb009955, 4-methoxy-3-bromo benzoic acid, 3-bromanissaure, pubchem2675, 2-bromo-4-carboxyanisole
PubChem CID 66836
IUPAC Name 3-bromo-4-methoxybenzoic acid
SMILES COC1=CC=C(C=C1Br)C([O-])=O

Specifications

Melting Point 217°C to 222°C
Odor Odorless
Quantity 5 g
Beilstein 2087585
Formula Weight 231.05
Percent Purity ≥98%
Chemical Name or Material 3-Bromo-4-methoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-768-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only