missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methoxybenzaldehyde, 98%

Catalog No. AAA1527406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1527406 5 g
AAA1527414 25 g
AAA1527422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1527406 Supplier Thermo Scientific Chemicals Supplier No. A1527406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34841-06-0 | C8H7BrO2 | 215.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34841-06-0
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00016599
InChI Key QMPNFQLVIGPNEI-UHFFFAOYSA-N
Synonym 3-bromo-p-anisaldehyde, benzaldehyde, 3-bromo-4-methoxy, 3-bromo-4-methoxy-benzaldehyde, 3-bromoanisaldehyde, 5-bromoanisaldehyde, 3-bromo-anisaldehyde, 5-bromo anisaldehyde, pubchem2643, acmc-209iat, 2-bromo-4-formylanisole
PubChem CID 98662
IUPAC Name 3-bromo-4-methoxybenzaldehyde
SMILES COC1=CC=C(C=O)C=C1Br

Specifications

Melting Point 49°C to 53°C
Boiling Point 108°C to 110°C (1 mmHg)
Quantity 5 g
Beilstein 1636939
Sensitivity Air Sensitive
Formula Weight 215.05
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-methoxybenzaldehyde
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 252-241-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only