missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methoxyaniline 96.0+%, TCI America™

Catalog No. B33201G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B33201G Supplier TCI America Supplier No. B33201G
Only null left

Chemical Identifiers

CAS 19056-41-8
Molecular Formula C7H8BrNO
Molecular Weight (g/mol) 202.051
MDL Number MFCD04094339
InChI Key NMUFTXMBONJQTC-UHFFFAOYSA-N
Synonym 4-Amino-2-bromoanisole
PubChem CID 961083
IUPAC Name 3-bromo-4-methoxyaniline
SMILES COC1=C(C=C(C=C1)N)Br

Specifications

Melting Point 62°C
Quantity 1 g
UN Number 2811
Formula Weight 202.05
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-methoxyaniline