missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4-hydroxybenzonitrile 95.0+%, TCI America™
$126.76 - $382.13
Chemical Identifiers
| CAS | 2315-86-8 |
|---|---|
| Molecular Formula | C7H4BrNO |
| Molecular Weight (g/mol) | 198.02 |
| MDL Number | MFCD00143096 |
| InChI Key | HLHNOIAOWQFNGW-UHFFFAOYSA-N |
| Synonym | 2-bromo-4-cyanophenol, 3-bromo-4-hydroxy-benzonitrile, benzonitrile, 3-bromo-4-hydroxy, 3-bromo-4-hydroxybenzenecarbonitrile, timtec-bb sbb005818, labotest-bb lt01143437, pubchem3780, intermediates-zcf02624, acmc-209g1r |
| PubChem CID | 75341 |
| IUPAC Name | 3-bromo-4-hydroxybenzonitrile |
| SMILES | OC1=CC=C(C=C1Br)C#N |
Chemical Identifiers
| 2315-86-8 | |
| 198.02 | |
| HLHNOIAOWQFNGW-UHFFFAOYSA-N | |
| 75341 | |
| OC1=CC=C(C=C1Br)C#N |
| C7H4BrNO | |
| MFCD00143096 | |
| 2-bromo-4-cyanophenol, 3-bromo-4-hydroxy-benzonitrile, benzonitrile, 3-bromo-4-hydroxy, 3-bromo-4-hydroxybenzenecarbonitrile, timtec-bb sbb005818, labotest-bb lt01143437, pubchem3780, intermediates-zcf02624, acmc-209g1r | |
| 3-bromo-4-hydroxybenzonitrile |
Specifications
| 2315-86-8 | |
| C7H4BrNO | |
| 5 g | |
| HLHNOIAOWQFNGW-UHFFFAOYSA-N | |
| 3-bromo-4-hydroxybenzonitrile | |
| 75341 | |
| ≥95.0% (GC) | |
| 3-Bromo-4-hydroxybenzonitrile |
| 156°C | |
| MFCD00143096 | |
| 2-bromo-4-cyanophenol, 3-bromo-4-hydroxy-benzonitrile, benzonitrile, 3-bromo-4-hydroxy, 3-bromo-4-hydroxybenzenecarbonitrile, timtec-bb sbb005818, labotest-bb lt01143437, pubchem3780, intermediates-zcf02624, acmc-209g1r | |
| OC1=CC=C(C=C1Br)C#N | |
| 198.02 | |
| 198.02 | |
| Crystalline Powder |
Safety and Handling
TSCA : No