missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-hydroxybenzonitrile, 98%

CAS: 2315-86-8 | C7H4BrNO | 198.02 g/mol

$86.86 - $284.22

Chemical Identifiers

CAS 2315-86-8
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.02
MDL Number MFCD00143096
InChI Key HLHNOIAOWQFNGW-UHFFFAOYSA-N
Synonym 2-bromo-4-cyanophenol, 3-bromo-4-hydroxy-benzonitrile, benzonitrile, 3-bromo-4-hydroxy, 3-bromo-4-hydroxybenzenecarbonitrile, timtec-bb sbb005818, labotest-bb lt01143437, pubchem3780, intermediates-zcf02624, acmc-209g1r
PubChem CID 75341
IUPAC Name 3-bromo-4-hydroxybenzonitrile
SMILES OC1=CC=C(C=C1Br)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1940006
View Documents
Thermo Scientific Chemicals
A1940006
5 g
Each for $86.86
Only null left
 
AAA1940014
View Documents
Thermo Scientific Chemicals
A1940014
25 g
Each for $284.22
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2315-86-8
198.02
HLHNOIAOWQFNGW-UHFFFAOYSA-N
75341
OC1=CC=C(C=C1Br)C#N
C7H4BrNO
MFCD00143096
2-bromo-4-cyanophenol, 3-bromo-4-hydroxy-benzonitrile, benzonitrile, 3-bromo-4-hydroxy, 3-bromo-4-hydroxybenzenecarbonitrile, timtec-bb sbb005818, labotest-bb lt01143437, pubchem3780, intermediates-zcf02624, acmc-209g1r
3-bromo-4-hydroxybenzonitrile

Specifications

2315-86-8
C7H4BrNO
5 g
2207020
HLHNOIAOWQFNGW-UHFFFAOYSA-N
3-bromo-4-hydroxybenzonitrile
75341
98%
156°C to 161°C
MFCD00143096
UN3439
2-bromo-4-cyanophenol, 3-bromo-4-hydroxy-benzonitrile, benzonitrile, 3-bromo-4-hydroxy, 3-bromo-4-hydroxybenzenecarbonitrile, timtec-bb sbb005818, labotest-bb lt01143437, pubchem3780, intermediates-zcf02624, acmc-209g1r
OC1=CC=C(C=C1Br)C#N
198.02
198.02
3-Bromo-4-hydroxybenzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 219-022-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only