missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-hydroxybenzonitrile, 98%

Catalog No. AAA1940006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1940006 5 g
AAA1940014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1940006 Supplier Thermo Scientific Chemicals Supplier No. A1940006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2315-86-8 | C7H4BrNO | 198.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2315-86-8
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.02
MDL Number MFCD00143096
InChI Key HLHNOIAOWQFNGW-UHFFFAOYSA-N
Synonym 2-bromo-4-cyanophenol, 3-bromo-4-hydroxy-benzonitrile, benzonitrile, 3-bromo-4-hydroxy, 3-bromo-4-hydroxybenzenecarbonitrile, timtec-bb sbb005818, labotest-bb lt01143437, pubchem3780, intermediates-zcf02624, acmc-209g1r
PubChem CID 75341
IUPAC Name 3-bromo-4-hydroxybenzonitrile
SMILES OC1=CC=C(C=C1Br)C#N

Specifications

Melting Point 156°C to 161°C
Quantity 5 g
UN Number UN3439
Beilstein 2207020
Formula Weight 198.02
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-hydroxybenzonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 219-022-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only