missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-hydroxybenzoic acid, 97%

Catalog No. AAB2133818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2133818 50 g
AAB2133809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2133818 Supplier Thermo Scientific Chemicals Supplier No. B2133818
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14348-41-5 | C7H5BrO3 | 217.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14348-41-5
Molecular Formula C7H5BrO3
Molecular Weight (g/mol) 217.02
MDL Number MFCD00017547
InChI Key XMEQDAIDOBVHEK-UHFFFAOYSA-N
Synonym 3-bromo-4-hydroxy-benzoic acid, benzoic acid, 3-bromo-4-hydroxy, acmc-20aix6, 3-bromo-4-hydroxybenzoicacid, 3-bromo-4-hydroxybenzoic acid, 3-bromo-4-hydroxybenzoicacidhydrate, 3-bromo-4-hydroxybenzoic acid 10g
PubChem CID 84368
IUPAC Name 3-bromo-4-hydroxybenzoic acid
SMILES OC(=O)C1=CC=C(O)C(Br)=C1

Specifications

Melting Point 176°C to 178°C
Quantity 50 g
Formula Weight 217.02
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-hydroxybenzoic acid
EINECSNumber 238-299-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only