missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4-fluorobenzyl Bromide 97.0+%, TCI America™
$91.33 - $290.79
Chemical Identifiers
| CAS | 78239-71-1 |
|---|---|
| Molecular Formula | C7H5Br2F |
| Molecular Weight (g/mol) | 267.92 |
| MDL Number | MFCD09037367 |
| InChI Key | ZRWSODQPUJMFRV-UHFFFAOYSA-N |
| PubChem CID | 12922710 |
| IUPAC Name | 2-bromo-4-(bromomethyl)-1-fluorobenzene |
| SMILES | FC1=C(Br)C=C(CBr)C=C1 |
Chemical Identifiers
| 78239-71-1 | |
| 267.92 | |
| ZRWSODQPUJMFRV-UHFFFAOYSA-N | |
| 2-bromo-4-(bromomethyl)-1-fluorobenzene |
| C7H5Br2F | |
| MFCD09037367 | |
| 12922710 | |
| FC1=C(Br)C=C(CBr)C=C1 |
Specifications
| 78239-71-1 | |
| C7H5Br2F | |
| 5 g | |
| 3-bromo-4-fluorobenzyl bromide, 2-bromo-4-bromomethyl-1-fluorobenzene, 3-bromo-4-fluorobenzylbromide, benzene, 2-bromo-4-bromomethyl-1-fluoro, 2-bromanyl-4-bromomethyl-1-fluoranyl-benzene, pubchem24015, acmc-209pdt, 3-bromo-4-fluoro-benzyl bromide, 2-bromo-4-bromomethyl-1-fluoro-benzene, benzene,2-bromo-4-bromomethyl-1-fluoro | |
| FC1=C(Br)C=C(CBr)C=C1 | |
| 267.92 | |
| 267.92 | |
| Crystalline Powder |
| 41°C | |
| MFCD09037367 | |
| 3261 | |
| ZRWSODQPUJMFRV-UHFFFAOYSA-N | |
| 2-bromo-4-(bromomethyl)-1-fluorobenzene | |
| 12922710 | |
| ≥97.0% (GC) | |
| 3-Bromo-4-fluorobenzyl Bromide |
Safety and Handling
TSCA : No