missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorobenzyl Bromide 97.0+%, TCI America™

Catalog No. B29755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29755G 5 g
B297525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29755G Supplier TCI America Supplier No. B29755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78239-71-1
Molecular Formula C7H5Br2F
Molecular Weight (g/mol) 267.92
MDL Number MFCD09037367
InChI Key ZRWSODQPUJMFRV-UHFFFAOYSA-N
PubChem CID 12922710
IUPAC Name 2-bromo-4-(bromomethyl)-1-fluorobenzene
SMILES FC1=C(Br)C=C(CBr)C=C1

Specifications

Melting Point 41°C
Quantity 5 g
UN Number 3261
Synonym 3-bromo-4-fluorobenzyl bromide, 2-bromo-4-bromomethyl-1-fluorobenzene, 3-bromo-4-fluorobenzylbromide, benzene, 2-bromo-4-bromomethyl-1-fluoro, 2-bromanyl-4-bromomethyl-1-fluoranyl-benzene, pubchem24015, acmc-209pdt, 3-bromo-4-fluoro-benzyl bromide, 2-bromo-4-bromomethyl-1-fluoro-benzene, benzene,2-bromo-4-bromomethyl-1-fluoro
Formula Weight 267.92
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-fluorobenzyl Bromide