missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4-fluorobenzyl Alcohol 98.0+%, TCI America™
$107.80 - $438.93
Chemical Identifiers
| CAS | 77771-03-0 |
|---|---|
| Molecular Formula | C7H6BrFO |
| Molecular Weight (g/mol) | 205.026 |
| MDL Number | MFCD00143093 |
| InChI Key | HNVVROFWONXXGO-UHFFFAOYSA-N |
| Synonym | 3-bromo-4-fluorobenzyl alcohol, 3-bromo-4-fluorophenyl methanol, 3-bromo-4-fluorobenzylalcohol, 3-bromo-4-fluoro-phenyl-methanol, 3-bromo-4-fluorophenyl methan-1-ol, pubchem4884, 4-fluoro-3-bromobenzyl alcohol, 3-bromo-4-fluoro-benzyl alcohol, 4-fluoro-3-bromo-benzyl alcohol, 3-bromo-4-fluoro benzyl alcohol |
| PubChem CID | 2773351 |
| IUPAC Name | (3-bromo-4-fluorophenyl)methanol |
| SMILES | C1=CC(=C(C=C1CO)Br)F |
Chemical Identifiers
| 77771-03-0 | |
| 205.026 | |
| HNVVROFWONXXGO-UHFFFAOYSA-N | |
| 2773351 | |
| C1=CC(=C(C=C1CO)Br)F |
| C7H6BrFO | |
| MFCD00143093 | |
| 3-bromo-4-fluorobenzyl alcohol, 3-bromo-4-fluorophenyl methanol, 3-bromo-4-fluorobenzylalcohol, 3-bromo-4-fluoro-phenyl-methanol, 3-bromo-4-fluorophenyl methan-1-ol, pubchem4884, 4-fluoro-3-bromobenzyl alcohol, 3-bromo-4-fluoro-benzyl alcohol, 4-fluoro-3-bromo-benzyl alcohol, 3-bromo-4-fluoro benzyl alcohol | |
| (3-bromo-4-fluorophenyl)methanol |
Specifications
| 77771-03-0 | |
| 90°C | |
| MFCD00143093 | |
| 3-bromo-4-fluorobenzyl alcohol, 3-bromo-4-fluorophenyl methanol, 3-bromo-4-fluorobenzylalcohol, 3-bromo-4-fluoro-phenyl-methanol, 3-bromo-4-fluorophenyl methan-1-ol, pubchem4884, 4-fluoro-3-bromobenzyl alcohol, 3-bromo-4-fluoro-benzyl alcohol, 4-fluoro-3-bromo-benzyl alcohol, 3-bromo-4-fluoro benzyl alcohol | |
| C1=CC(=C(C=C1CO)Br)F | |
| 205.026 | |
| 205.03 | |
| Liquid |
| Yellow | |
| C7H6BrFO | |
| 5 g | |
| HNVVROFWONXXGO-UHFFFAOYSA-N | |
| (3-bromo-4-fluorophenyl)methanol | |
| 2773351 | |
| ≥98.0% (GC) | |
| 3-Bromo-4-fluorobenzyl Alcohol |
Safety and Handling
TSCA : No