missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorobenzyl Alcohol 98.0+%, TCI America™

Catalog No. B46565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46565G 5 g
B465625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46565G Supplier TCI America Supplier No. B46565G

Chemical Identifiers

CAS 77771-03-0
Molecular Formula C7H6BrFO
Molecular Weight (g/mol) 205.026
MDL Number MFCD00143093
InChI Key HNVVROFWONXXGO-UHFFFAOYSA-N
Synonym 3-bromo-4-fluorobenzyl alcohol, 3-bromo-4-fluorophenyl methanol, 3-bromo-4-fluorobenzylalcohol, 3-bromo-4-fluoro-phenyl-methanol, 3-bromo-4-fluorophenyl methan-1-ol, pubchem4884, 4-fluoro-3-bromobenzyl alcohol, 3-bromo-4-fluoro-benzyl alcohol, 4-fluoro-3-bromo-benzyl alcohol, 3-bromo-4-fluoro benzyl alcohol
PubChem CID 2773351
IUPAC Name (3-bromo-4-fluorophenyl)methanol
SMILES C1=CC(=C(C=C1CO)Br)F

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
Formula Weight 205.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-4-fluorobenzyl Alcohol