missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4-fluorobenzyl alcohol, 97%
CAS: 77771-03-0 | C7H6BrFO | 205.026 g/mol
$59.14 - $195.95
Chemical Identifiers
| CAS | 77771-03-0 |
|---|---|
| Molecular Formula | C7H6BrFO |
| Molecular Weight (g/mol) | 205.026 |
| MDL Number | MFCD00143093 |
| InChI Key | HNVVROFWONXXGO-UHFFFAOYSA-N |
| Synonym | 3-bromo-4-fluorobenzyl alcohol, 3-bromo-4-fluorophenyl methanol, 3-bromo-4-fluorobenzylalcohol, 3-bromo-4-fluoro-phenyl-methanol, 3-bromo-4-fluorophenyl methan-1-ol, pubchem4884, 4-fluoro-3-bromobenzyl alcohol, 3-bromo-4-fluoro-benzyl alcohol, 4-fluoro-3-bromo-benzyl alcohol, 3-bromo-4-fluoro benzyl alcohol |
| PubChem CID | 2773351 |
| IUPAC Name | (3-bromo-4-fluorophenyl)methanol |
| SMILES | C1=CC(=C(C=C1CO)Br)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3060406
|
Thermo Scientific Chemicals
H3060406 |
5 g |
Each for $59.14
|
|
|||||
|
AAH3060414
|
Thermo Scientific Chemicals
H3060414 |
25 g |
Each for $195.95
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 77771-03-0 | |
| 205.026 | |
| HNVVROFWONXXGO-UHFFFAOYSA-N | |
| 2773351 | |
| C1=CC(=C(C=C1CO)Br)F |
| C7H6BrFO | |
| MFCD00143093 | |
| 3-bromo-4-fluorobenzyl alcohol, 3-bromo-4-fluorophenyl methanol, 3-bromo-4-fluorobenzylalcohol, 3-bromo-4-fluoro-phenyl-methanol, 3-bromo-4-fluorophenyl methan-1-ol, pubchem4884, 4-fluoro-3-bromobenzyl alcohol, 3-bromo-4-fluoro-benzyl alcohol, 4-fluoro-3-bromo-benzyl alcohol, 3-bromo-4-fluoro benzyl alcohol | |
| (3-bromo-4-fluorophenyl)methanol |
Specifications
| 77771-03-0 | |
| C7H6BrFO | |
| 5 g | |
| HNVVROFWONXXGO-UHFFFAOYSA-N | |
| (3-bromo-4-fluorophenyl)methanol | |
| 2773351 | |
| 97% |
| 80°C (0.5 mmHg) | |
| MFCD00143093 | |
| 3-bromo-4-fluorobenzyl alcohol, 3-bromo-4-fluorophenyl methanol, 3-bromo-4-fluorobenzylalcohol, 3-bromo-4-fluoro-phenyl-methanol, 3-bromo-4-fluorophenyl methan-1-ol, pubchem4884, 4-fluoro-3-bromobenzyl alcohol, 3-bromo-4-fluoro-benzyl alcohol, 4-fluoro-3-bromo-benzyl alcohol, 3-bromo-4-fluoro benzyl alcohol | |
| C1=CC(=C(C=C1CO)Br)F | |
| 205.026 | |
| 205.03 | |
| 3-Bromo-4-fluorobenzyl alcohol |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only