missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorobenzyl alcohol, 97%

Catalog No. AAH3060414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3060414 25 g
AAH3060406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3060414 Supplier Thermo Scientific Chemicals Supplier No. H3060414

CAS: 77771-03-0 | C7H6BrFO | 205.026 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 77771-03-0
Molecular Formula C7H6BrFO
Molecular Weight (g/mol) 205.026
MDL Number MFCD00143093
InChI Key HNVVROFWONXXGO-UHFFFAOYSA-N
Synonym 3-bromo-4-fluorobenzyl alcohol, 3-bromo-4-fluorophenyl methanol, 3-bromo-4-fluorobenzylalcohol, 3-bromo-4-fluoro-phenyl-methanol, 3-bromo-4-fluorophenyl methan-1-ol, pubchem4884, 4-fluoro-3-bromobenzyl alcohol, 3-bromo-4-fluoro-benzyl alcohol, 4-fluoro-3-bromo-benzyl alcohol, 3-bromo-4-fluoro benzyl alcohol
PubChem CID 2773351
IUPAC Name (3-bromo-4-fluorophenyl)methanol
SMILES C1=CC(=C(C=C1CO)Br)F

Specifications

Boiling Point 80°C (0.5 mmHg)
Quantity 25 g
Formula Weight 205.03
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-fluorobenzyl alcohol
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only