missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorobenzotrifluoride 96.0+%, TCI America™

Catalog No. B14215G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14215G 5 g
B142125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14215G Supplier TCI America Supplier No. B14215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68322-84-9
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 243.00
MDL Number MFCD00040938
InChI Key RZJOIMPUMMQKFR-UHFFFAOYSA-N
Synonym 3-bromo-4-fluorobenzotrifluoride, 2-bromo-1-fluoro-4-trifluoromethyl benzene, 2-fluoro-5-trifluoromethyl bromobenzene, 3-bromo-alpha,alpha,alpha,4-tetrafluorotoluene, benzene, 2-bromo-1-fluoro-4-trifluoromethyl, 1-bromo-2-fluoro-5-trifluoromethyl benzene, 3-bromo-alpha,alpha,alpha-4-tetrafluorotoluene, pubchem1639, 3-bromo-4-fluoro trifluoromethyl benzene
PubChem CID 144295
IUPAC Name 2-bromo-1-fluoro-4-(trifluoromethyl)benzene
SMILES FC1=CC=C(C=C1Br)C(F)(F)F

Specifications

Color Yellow
Boiling Point 60°C
Quantity 5 g
Formula Weight 243.00
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-4-fluorobenzotrifluoride
TSCA No
Recommended Storage Refrigerator