missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4-fluorobenzoic Acid 96.0+%, TCI America™
$50.33 - $145.40
Chemical Identifiers
| CAS | 1007-16-5 |
|---|---|
| Molecular Formula | C7H3BrFO2 |
| Molecular Weight (g/mol) | 218.00 |
| MDL Number | MFCD00042463 |
| InChI Key | ONELILMJNOWXSA-UHFFFAOYSA-M |
| Synonym | 3-bromo-4-fluoro-benzoic acid, 4-fluoro-3-bromo benzoic acid, 3-bromo-4-fluorobenzoicacid, rarechem al bo 0604, buttpark 20\01-58, benzoic acid, 3-bromo-4-fluoro, 3-bromo-4-fluorbenzoic acid, pubchem1320, 3-bromo-4-fiuorobenzoic acid, 5-bromo-4-fluorobenzoic acid |
| PubChem CID | 70509 |
| IUPAC Name | 3-bromo-4-fluorobenzoate |
| SMILES | [O-]C(=O)C1=CC=C(F)C(Br)=C1 |
Chemical Identifiers
| 1007-16-5 | |
| 218.00 | |
| ONELILMJNOWXSA-UHFFFAOYSA-M | |
| 70509 | |
| [O-]C(=O)C1=CC=C(F)C(Br)=C1 |
| C7H3BrFO2 | |
| MFCD00042463 | |
| 3-bromo-4-fluoro-benzoic acid, 4-fluoro-3-bromo benzoic acid, 3-bromo-4-fluorobenzoicacid, rarechem al bo 0604, buttpark 20\01-58, benzoic acid, 3-bromo-4-fluoro, 3-bromo-4-fluorbenzoic acid, pubchem1320, 3-bromo-4-fiuorobenzoic acid, 5-bromo-4-fluorobenzoic acid | |
| 3-bromo-4-fluorobenzoate |
Specifications
| 1007-16-5 | |
| White | |
| MFCD00042463 | |
| 3-bromo-4-fluoro-benzoic acid, 4-fluoro-3-bromo benzoic acid, 3-bromo-4-fluorobenzoicacid, rarechem al bo 0604, buttpark 20\01-58, benzoic acid, 3-bromo-4-fluoro, 3-bromo-4-fluorbenzoic acid, pubchem1320, 3-bromo-4-fiuorobenzoic acid, 5-bromo-4-fluorobenzoic acid | |
| [O-]C(=O)C1=CC=C(F)C(Br)=C1 | |
| 218.00 | |
| 219.01 | |
| Crystalline Powder |
| 139°C | |
| C7H3BrFO2 | |
| 1 g | |
| ONELILMJNOWXSA-UHFFFAOYSA-M | |
| 3-bromo-4-fluorobenzoate | |
| 70509 | |
| ≥96.0% (T) | |
| 3-Bromo-4-fluorobenzoic Acid |
Safety and Handling
TSCA : No