missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4-fluorobenzaldehyde 97.0+%, TCI America™
$67.11 - $201.32
Chemical Identifiers
| CAS | 77771-02-9 |
|---|---|
| Molecular Formula | C7H4BrFO |
| Molecular Weight (g/mol) | 203.01 |
| MDL Number | MFCD00042186 |
| InChI Key | FAHZIKXYYRGSHF-UHFFFAOYSA-N |
| Synonym | 4-fluoro-3-bromobenzaldehyde, benzaldehyde, 3-bromo-4-fluoro, 3-bromo-4-fluoro-benzaldehyde, 3-bromo-4-fluoro benzaldehyde, pubchem2878, acmc-209pbm, 3-bromo4fluorobenzaldehyde, 3-bromo-4fluorobenzaldehyde, 3-bromo4-fluorobenzaldehyde, 3bromo-4-fluorobenzaldehyde |
| PubChem CID | 173604 |
| IUPAC Name | 3-bromo-4-fluorobenzaldehyde |
| SMILES | C1=CC(=C(C=C1C=O)Br)F |
Chemical Identifiers
| 77771-02-9 | |
| 203.01 | |
| FAHZIKXYYRGSHF-UHFFFAOYSA-N | |
| 173604 | |
| C1=CC(=C(C=C1C=O)Br)F |
| C7H4BrFO | |
| MFCD00042186 | |
| 4-fluoro-3-bromobenzaldehyde, benzaldehyde, 3-bromo-4-fluoro, 3-bromo-4-fluoro-benzaldehyde, 3-bromo-4-fluoro benzaldehyde, pubchem2878, acmc-209pbm, 3-bromo4fluorobenzaldehyde, 3-bromo-4fluorobenzaldehyde, 3-bromo4-fluorobenzaldehyde, 3bromo-4-fluorobenzaldehyde | |
| 3-bromo-4-fluorobenzaldehyde |
Specifications
| 77771-02-9 | |
| White-Yellow | |
| C7H4BrFO | |
| 5 g | |
| FAHZIKXYYRGSHF-UHFFFAOYSA-N | |
| 3-bromo-4-fluorobenzaldehyde | |
| 173604 | |
| ≥97.0% (GC) | |
| 3-Bromo-4-fluorobenzaldehyde |
| 33°C | |
| 139°C | |
| MFCD00042186 | |
| 4-fluoro-3-bromobenzaldehyde, benzaldehyde, 3-bromo-4-fluoro, 3-bromo-4-fluoro-benzaldehyde, 3-bromo-4-fluoro benzaldehyde, pubchem2878, acmc-209pbm, 3-bromo4fluorobenzaldehyde, 3-bromo-4fluorobenzaldehyde, 3-bromo4-fluorobenzaldehyde, 3bromo-4-fluorobenzaldehyde | |
| C1=CC(=C(C=C1C=O)Br)F | |
| 203.01 | |
| 203.01 | |
| Lumps |
Safety and Handling
TSCA : Yes