missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorobenzaldehyde, 98%

Catalog No. AAA1519814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1519814 25 g
AAA1519806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1519814 Supplier Thermo Scientific Chemicals Supplier No. A1519814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 77771-02-9 | C7H4BrFO | 203.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 77771-02-9
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD00042186
InChI Key FAHZIKXYYRGSHF-UHFFFAOYSA-N
Synonym 4-fluoro-3-bromobenzaldehyde, benzaldehyde, 3-bromo-4-fluoro, 3-bromo-4-fluoro-benzaldehyde, 3-bromo-4-fluoro benzaldehyde, pubchem2878, acmc-209pbm, 3-bromo4fluorobenzaldehyde, 3-bromo-4fluorobenzaldehyde, 3-bromo4-fluorobenzaldehyde, 3bromo-4-fluorobenzaldehyde
PubChem CID 173604
IUPAC Name 3-bromo-4-fluorobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)Br)F

Specifications

Melting Point 29°C to 33°C
Boiling Point 138°C to 139°C (2.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.574
Quantity 25 g
Beilstein 5806226
Sensitivity Air Sensitive
Formula Weight 203.01
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-fluorobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 278-764-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only