missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluoroaniline 97.0+%, TCI America™

Catalog No. B29575G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B29575G Supplier TCI America Supplier No. B29575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 656-64-4
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD03788559
InChI Key KOWPUNQBGWIERF-UHFFFAOYSA-N
Synonym 3-bromo-4-fluoro aniline, benzenamine, 3-bromo-4-fluoro, 3-bromo-4-fluorobenzenamine, 3-bromo-4-fluorophenylamine, 3-bromo-4-fluoro-aniline, 3-bromo-4-fluoro-phenylamine, pubchem2924, acmc-209nrn, 3-bromo-4-fluoraniline, 3-bromo 4-fluoro-aniline
PubChem CID 2779286
IUPAC Name 3-bromo-4-fluoroaniline
SMILES C1=CC(=C(C=C1N)Br)F

Specifications

Color White-Yellow
Boiling Point 124°C
Quantity 5 g
Formula Weight 190.02
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Bromo-4-fluoroaniline