missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluoroaniline, 98%

Catalog No. AAB2561006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2561006 5 g
AAB2561014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2561006 Supplier Thermo Scientific Chemicals Supplier No. B2561006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 656-64-4 | C6H5BrFN | 190.015 g/mol

3-Bromo-4-fluoroaniline is used in organic synthesis as an intermediate of agrochemicals and active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-fluoroaniline is used in organic synthesis as an intermediate of agrochemicals and active pharmaceutical ingredient.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.

Chemical Identifiers

CAS 656-64-4
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD03788559
InChI Key KOWPUNQBGWIERF-UHFFFAOYSA-N
Synonym 3-bromo-4-fluoro aniline, benzenamine, 3-bromo-4-fluoro, 3-bromo-4-fluorobenzenamine, 3-bromo-4-fluorophenylamine, 3-bromo-4-fluoro-aniline, 3-bromo-4-fluoro-phenylamine, pubchem2924, acmc-209nrn, 3-bromo-4-fluoraniline, 3-bromo 4-fluoro-aniline
PubChem CID 2779286
IUPAC Name 3-bromo-4-fluoroaniline
SMILES C1=CC(=C(C=C1N)Br)F

Specifications

Melting Point 34°C to 36°C
Boiling Point 235°C
Quantity 5 g
UN Number UN2811
Sensitivity Air sensitive
Solubility Information Insoluble in water.
Formula Weight 190.01
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-fluoroaniline
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only