missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3'-Bromo-4'-fluoroacetophenone 96.0+%, TCI America™
Supplier: TCI America B201425G
Specifications
| 3′-Bromo-4′-fluoroacetophenone | |
| 56°C | |
| 240°C | |
| MFCD00042466 | |
| 3'-bromo-4'-fluoroacetophenone, 3-bromo-4-fluoroacetophenone, 1-3-bromo-4-fluorophenyl ethanone, 1-3-bromo-4-fluorophenyl ethan-1-one, ethanone, 1-3-bromo-4-fluorophenyl, 1-3-bromo-4-fluoro-phenyl-ethanone, 1-acetyl-3-bromo-4-fluorobenzene, pubchem4100, acmc-1bol8, ksc499c8t | |
| CC(=O)C1=CC=C(F)C(Br)=C1 | |
| 217.04 | |
| 217.04 | |
| Crystalline Powder |
| 1007-15-4 | |
| White-Yellow | |
| C8H6BrFO | |
| 25 g | |
| SZDWTGAORQQQGY-UHFFFAOYSA-N | |
| 1-(3-bromo-4-fluorophenyl)ethan-1-one | |
| 70508 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 1007-15-4 | |
| 217.04 | |
| SZDWTGAORQQQGY-UHFFFAOYSA-N | |
| 70508 | |
| CC(=O)C1=CC=C(F)C(Br)=C1 |
| C8H6BrFO | |
| MFCD00042466 | |
| 3'-bromo-4'-fluoroacetophenone, 3-bromo-4-fluoroacetophenone, 1-3-bromo-4-fluorophenyl ethanone, 1-3-bromo-4-fluorophenyl ethan-1-one, ethanone, 1-3-bromo-4-fluorophenyl, 1-3-bromo-4-fluoro-phenyl-ethanone, 1-acetyl-3-bromo-4-fluorobenzene, pubchem4100, acmc-1bol8, ksc499c8t | |
| 1-(3-bromo-4-fluorophenyl)ethan-1-one |
Safety and Handling
TSCA : No