accessibility menu, dialog, popup

UserName

CAS RN 1007-15-4

IUPAC Name: 1-(3-bromo-4-fluorophenyl)ethan-1-one
Synonyms: 1-(3-bromo-4-fluorophenyl)ethanone
Molecular Weight (g/mol): 217.04
Molecular Formula: C8H6BrFO
InChi Key: SZDWTGAORQQQGY-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(F)C(Br)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)