missing translation for 'onlineSavingsMsg'
Learn More

3'-Bromo-4'-fluoroacetophenone 96.0+%, TCI America™

Catalog No. B20145G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20145G 5 g
B201425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20145G Supplier TCI America Supplier No. B20145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1007-15-4
Molecular Formula C8H6BrFO
Molecular Weight (g/mol) 217.04
MDL Number MFCD00042466
InChI Key SZDWTGAORQQQGY-UHFFFAOYSA-N
Synonym 3'-bromo-4'-fluoroacetophenone, 3-bromo-4-fluoroacetophenone, 1-3-bromo-4-fluorophenyl ethanone, 1-3-bromo-4-fluorophenyl ethan-1-one, ethanone, 1-3-bromo-4-fluorophenyl, 1-3-bromo-4-fluoro-phenyl-ethanone, 1-acetyl-3-bromo-4-fluorobenzene, pubchem4100, acmc-1bol8, ksc499c8t
PubChem CID 70508
IUPAC Name 1-(3-bromo-4-fluorophenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(F)C(Br)=C1

Specifications

Melting Point 56°C
Color White-Yellow
Boiling Point 240°C
Quantity 5 g
Formula Weight 217.04
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Bromo-4′-fluoroacetophenone