missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4-chloronitrobenzene 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 58°C |
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 236.45 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Bromo-4-chloronitrobenzene |
Safety and Handling
| TSCA | No |