missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-chlorobenzotrifluoride 97.0+%, TCI America™

Catalog No. B274325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B274325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B274325G Supplier TCI America Supplier No. B274325G
Only null left

Chemical Identifiers

CAS 454-78-4
Molecular Formula C7H3BrClF3
Molecular Weight (g/mol) 259.45
InChI Key APSISOSWYXCEQX-UHFFFAOYSA-N
Synonym 2-bromo-1-chloro-4-trifluoromethyl benzene, 3-bromo-4-chlorobenzotrifluoride, benzene, 2-bromo-1-chloro-4-trifluoromethyl, 3-bromo-4-chloro-alpha,alpha,alpha-trifluorotoluene, 4-chloro-3-bromobenzotrifluoride, 3-bromo-4-chloro-a,a,a-trifluorotoluene, pubchem4837, ksc915a8j, apsisoswyxceqx-uhfffaoysa, 3-bromo-4-chloro-benzotrifluoride
PubChem CID 67988
IUPAC Name 2-bromo-1-chloro-4-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)Br)Cl

Specifications

Melting Point -22°C
Color Yellow
Boiling Point 190°C
Quantity 25 g
Formula Weight 259.45
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-4-chlorobenzotrifluoride