missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-chlorobenzoic Acid 98.0+%, TCI America™

Catalog No. B33085G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33085G 5 g
B33081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33085G Supplier TCI America Supplier No. B33085G

Chemical Identifiers

CAS 42860-10-6
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.46
MDL Number MFCD00079706
InChI Key NLEPZGNUPNMRGF-UHFFFAOYSA-N
Synonym 3-bromo-4-chloro-benzoic acid, benzoic acid, 3-bromo-4-chloro, 4-chloro-3-bromobenzoic acid, pubchem3585, acmc-1cufe, 3-bromo-4-chlorobenzoicacid, ksc235m9l, timtec-bb sbb017622, zerenex e/9071912
PubChem CID 2735544
IUPAC Name 3-bromo-4-chlorobenzoic acid
SMILES OC(=O)C1=CC(Br)=C(Cl)C=C1

Specifications

Melting Point 220°C
Color White-Yellow
Quantity 5 g
Formula Weight 235.46
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-chlorobenzoic Acid