missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-2-nitroanisole 98.0+%, TCI America™
$63.52 - $256.05
Chemical Identifiers
| CAS | 500298-30-6 |
|---|---|
| Molecular Formula | C7H6BrNO3 |
| Molecular Weight (g/mol) | 232.03 |
| MDL Number | MFCD11101044 |
| InChI Key | ORSPVBUIPTYLLO-UHFFFAOYSA-N |
| Synonym | 1-Bromo-3-methoxy-2-nitrobenzene |
| PubChem CID | 258734 |
| IUPAC Name | 1-bromo-3-methoxy-2-nitrobenzene |
| SMILES | COC1=C(C(Br)=CC=C1)[N+]([O-])=O |
Chemical Identifiers
| 500298-30-6 | |
| 232.03 | |
| ORSPVBUIPTYLLO-UHFFFAOYSA-N | |
| 258734 | |
| COC1=C(C(Br)=CC=C1)[N+]([O-])=O |
| C7H6BrNO3 | |
| MFCD11101044 | |
| 1-Bromo-3-methoxy-2-nitrobenzene | |
| 1-bromo-3-methoxy-2-nitrobenzene |
Specifications
| 500298-30-6 | |
| 131°C | |
| MFCD11101044 | |
| 1-Bromo-3-methoxy-2-nitrobenzene | |
| COC1=C(C(Br)=CC=C1)[N+]([O-])=O | |
| 232.03 | |
| 232.03 | |
| Crystalline Powder |
| 74°C | |
| C7H6BrNO3 | |
| 1 g | |
| ORSPVBUIPTYLLO-UHFFFAOYSA-N | |
| 1-bromo-3-methoxy-2-nitrobenzene | |
| 258734 | |
| ≥98.0% (GC) | |
| 3-Bromo-2-nitroanisole |
Safety and Handling
TSCA : No