missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-nitroanisole 98.0+%, TCI America™

Catalog No. B34861G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34861G 1 g
B34865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34861G Supplier TCI America Supplier No. B34861G
Only null left

Chemical Identifiers

CAS 500298-30-6
Molecular Formula C7H6BrNO3
Molecular Weight (g/mol) 232.03
MDL Number MFCD11101044
InChI Key ORSPVBUIPTYLLO-UHFFFAOYSA-N
Synonym 1-Bromo-3-methoxy-2-nitrobenzene
PubChem CID 258734
IUPAC Name 1-bromo-3-methoxy-2-nitrobenzene
SMILES COC1=C(C(Br)=CC=C1)[N+]([O-])=O

Specifications

Melting Point 74°C
Boiling Point 131°C
Quantity 1 g
Formula Weight 232.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-2-nitroanisole