missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-2-methylpyridine 98.0+%, TCI America™
$74.56 - $227.41
Chemical Identifiers
| CAS | 38749-79-0 |
|---|---|
| Molecular Formula | C6H6BrN |
| Molecular Weight (g/mol) | 172.03 |
| MDL Number | MFCD00191224 |
| InChI Key | AIPWPTPHMIYYOX-UHFFFAOYSA-N |
| Synonym | 3-bromo-2-picoline, 2-methyl-3-bromopyridine, 3-bromo-2-methyl-pyridine, pyridine, 3-bromo-2-methyl, 2-methyl-3-bromo pyridine, pubchem1220, 3-brom-2-methylpyridin, acmc-209j1t, 3-bromo-2-methyl pyridine, ksc497m1l |
| PubChem CID | 1201003 |
| IUPAC Name | 3-bromo-2-methylpyridine |
| SMILES | CC1=C(Br)C=CC=N1 |
Chemical Identifiers
| 38749-79-0 | |
| 172.03 | |
| AIPWPTPHMIYYOX-UHFFFAOYSA-N | |
| 1201003 | |
| CC1=C(Br)C=CC=N1 |
| C6H6BrN | |
| MFCD00191224 | |
| 3-bromo-2-picoline, 2-methyl-3-bromopyridine, 3-bromo-2-methyl-pyridine, pyridine, 3-bromo-2-methyl, 2-methyl-3-bromo pyridine, pubchem1220, 3-brom-2-methylpyridin, acmc-209j1t, 3-bromo-2-methyl pyridine, ksc497m1l | |
| 3-bromo-2-methylpyridine |
Specifications
| 38749-79-0 | |
| 76°C | |
| MFCD00191224 | |
| 3-bromo-2-picoline, 2-methyl-3-bromopyridine, 3-bromo-2-methyl-pyridine, pyridine, 3-bromo-2-methyl, 2-methyl-3-bromo pyridine, pubchem1220, 3-brom-2-methylpyridin, acmc-209j1t, 3-bromo-2-methyl pyridine, ksc497m1l | |
| CC1=C(Br)C=CC=N1 | |
| 172.03 | |
| 172.03 | |
| Liquid |
| Yellow | |
| C6H6BrN | |
| 1 g | |
| AIPWPTPHMIYYOX-UHFFFAOYSA-N | |
| 3-bromo-2-methylpyridine | |
| 1201003 | |
| ≥98.0% (GC) | |
| 3-Bromo-2-methylpyridine |
Safety and Handling
TSCA : No