accessibility menu, dialog, popup

UserName

CH3NBr

CAS RN 38749-79-0

IUPAC Name: 3-bromo-2-methylpyridine
Synonyms: 3-bromo-2-methylpyridine
Molecular Weight (g/mol): 172.03
Molecular Formula: C6H6BrN
InChi Key: AIPWPTPHMIYYOX-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=CC=N1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)