missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-methylpyridine 98.0+%, TCI America™

Catalog No. B29625G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29625G 5 g
B29621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29625G Supplier TCI America Supplier No. B29625G

Chemical Identifiers

CAS 38749-79-0
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD00191224
InChI Key AIPWPTPHMIYYOX-UHFFFAOYSA-N
Synonym 3-bromo-2-picoline, 2-methyl-3-bromopyridine, 3-bromo-2-methyl-pyridine, pyridine, 3-bromo-2-methyl, 2-methyl-3-bromo pyridine, pubchem1220, 3-brom-2-methylpyridin, acmc-209j1t, 3-bromo-2-methyl pyridine, ksc497m1l
PubChem CID 1201003
IUPAC Name 3-bromo-2-methylpyridine
SMILES CC1=C(Br)C=CC=N1

Specifications

Color Yellow
Boiling Point 76°C
Quantity 5 g
Formula Weight 172.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-2-methylpyridine