missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-methylpyridine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 38749-79-0
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD00191224
InChI Key AIPWPTPHMIYYOX-UHFFFAOYSA-N
Synonym 3-bromo-2-picoline, 2-methyl-3-bromopyridine, 3-bromo-2-methyl-pyridine, pyridine, 3-bromo-2-methyl, 2-methyl-3-bromo pyridine, pubchem1220, 3-brom-2-methylpyridin, acmc-209j1t, 3-bromo-2-methyl pyridine, ksc497m1l
PubChem CID 1201003
IUPAC Name 3-bromo-2-methylpyridine
SMILES CC1=C(Br)C=CC=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC436860010
SDS View Documents
Thermo Scientific Chemicals
436860010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC436860050
SDS View Documents
Thermo Scientific Chemicals
436860050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

38749-79-0
172.03
AIPWPTPHMIYYOX-UHFFFAOYSA-N
1201003
CC1=C(Br)C=CC=N1
C6H6BrN
MFCD00191224
3-bromo-2-picoline, 2-methyl-3-bromopyridine, 3-bromo-2-methyl-pyridine, pyridine, 3-bromo-2-methyl, 2-methyl-3-bromo pyridine, pubchem1220, 3-brom-2-methylpyridin, acmc-209j1t, 3-bromo-2-methyl pyridine, ksc497m1l
3-bromo-2-methylpyridine

Specifications

38749-79-0
100.0
65°C to 67°C (10.0 Torr)
Authentic
Glass bottle
MFCD00191224
1.495
AIPWPTPHMIYYOX-UHFFFAOYSA-N
3-bromo-2-methylpyridine
1201003
97%
96.0
1.4950g/mL
79°C
96% min. (GC)
C6H6BrN
1g
3-bromo-2-picoline, 2-methyl-3-bromopyridine, 3-bromo-2-methyl-pyridine, pyridine, 3-bromo-2-methyl, 2-methyl-3-bromo pyridine, pubchem1220, 3-brom-2-methylpyridin, acmc-209j1t, 3-bromo-2-methyl pyridine, ksc497m1l
CC1=C(Br)C=CC=N1
172.03
172.02
3-Bromo-2-methylpyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only