missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-methylpropene, 97%

Catalog No. p-4089598
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427650050 5 mL Glass bottle
AC427650250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC427650050 Supplier Thermo Scientific Chemicals Supplier No. 427650050

CAS: 1458-98-6 | C4H7Br | 135.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1458-98-6
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135.00
MDL Number MFCD00134155
InChI Key USEGQJLHQSTGHW-UHFFFAOYSA-N
Synonym 3-bromo-2-methylpropene, 3-bromo-2-methyl-1-propene, methallyl bromide, 2-bromomethyl prop-1-ene, 2-methylallyl bromide, 1-propene, 3-bromo-2-methyl, 3-bromo-2-methyl-prop-1-ene, 3-bromo-2-methylpropene, stabilized with hydroquinone, methallylbromide, methylallyl bromide
PubChem CID 357785
IUPAC Name 3-bromo-2-methylprop-1-ene
SMILES CC(=C)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.3390g/mL
Boiling Point 94.0°C to 95.0°C
Flash Point 7°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH2=C(CH3)CH2Br
Quantity 5 mL
Specific Gravity 1.339
Formula Weight 135
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-methylpropene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away fr

RUO – Research Use Only