missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-2-methylbenzoic Acid 98.0+%, TCI America™
Supplier: TCI America B32215G
Specifications
| 3-Bromo-2-methylbenzoic Acid | |
| 154°C | |
| C8H7BrO2 | |
| 5 g | |
| BJGKVCKGUBYULR-UHFFFAOYSA-N | |
| 3-bromo-2-methylbenzoic acid | |
| 2735590 | |
| ≥98.0% (GC,T) |
| 76006-33-2 | |
| White-Yellow | |
| MFCD00270097 | |
| 3-bromo-o-toluic acid, 2-methyl-3-bromobenzoic acid, 3-bromo-2-methyl-benzoic acid, benzoic acid, 3-bromo-2-methyl, pubchem4717, bromo-methylbenzoic acid, 2-bromo-6-carboxytoluene, 3-bromo-2-methybenzoic acid, 3-bromo-2-methylbenzoicacid, ksc377a4p | |
| CC1=C(C=CC=C1Br)C(O)=O | |
| 215.05 | |
| 215.05 | |
| Crystalline Powder |
Chemical Identifiers
| 76006-33-2 | |
| 215.05 | |
| BJGKVCKGUBYULR-UHFFFAOYSA-N | |
| 2735590 | |
| CC1=C(C=CC=C1Br)C(O)=O |
| C8H7BrO2 | |
| MFCD00270097 | |
| 3-bromo-o-toluic acid, 2-methyl-3-bromobenzoic acid, 3-bromo-2-methyl-benzoic acid, benzoic acid, 3-bromo-2-methyl, pubchem4717, bromo-methylbenzoic acid, 2-bromo-6-carboxytoluene, 3-bromo-2-methybenzoic acid, 3-bromo-2-methylbenzoicacid, ksc377a4p | |
| 3-bromo-2-methylbenzoic acid |
Safety and Handling
TSCA : No