missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-methylbenzoic acid, 97%

Catalog No. p-7036946
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5543603 1 g
AAH5543606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5543603 Supplier Thermo Scientific Chemicals Supplier No. H5543603

CAS: 76006-33-2 | C8H7BrO2 | 215.05 g/mol

3-Bromo-2-methylbenzoic Acid is a disubstituted benzoic acid used in the preparation of various biologically active compounds such as α-2 adrenoceptor agonists, Smoothened receptor anatogonists and HIV-1 entry inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-2-methylbenzoic Acid is a disubstituted benzoic acid used in the preparation of various biologically active compounds such as α-2 adrenoceptor agonists, Smoothened receptor anatogonists and HIV-1 entry inhibitors.

Solubility
Soluble in chloroform, methanol

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.

Chemical Identifiers

CAS 76006-33-2
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00270097
InChI Key BJGKVCKGUBYULR-UHFFFAOYSA-N
Synonym 3-bromo-o-toluic acid, 2-methyl-3-bromobenzoic acid, 3-bromo-2-methyl-benzoic acid, benzoic acid, 3-bromo-2-methyl, pubchem4717, bromo-methylbenzoic acid, 2-bromo-6-carboxytoluene, 3-bromo-2-methybenzoic acid, 3-bromo-2-methylbenzoicacid, ksc377a4p
PubChem CID 2735590
IUPAC Name 3-bromo-2-methylbenzoic acid
SMILES CC1=C(C=CC=C1Br)C(O)=O

Specifications

Melting Point 152°C to 156°C
Quantity 1 g
Solubility Information Soluble in chloroform,methanol
Formula Weight 215.05
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-methylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only