missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-methylaniline 98.0+%, TCI America™

Catalog No. B34825G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34825G 5 g
B348225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34825G Supplier TCI America Supplier No. B34825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 55289-36-6
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.052
MDL Number MFCD00051579
InChI Key IILVSKMKMOJHMA-UHFFFAOYSA-N
Synonym 2-amino-6-bromotoluene, 3-bromo-o-toluidine, benzenamine, 3-bromo-2-methyl, 3-bromo-2-methylbenzenamine, 2-bromo-6-aminotoluene, 3-bromo-2-methyl-phenylamine, 3-bromo-2-methyl-aniline, 3-bromo-2-methylphenylamine, 3-bromo-2-methyl-benzenamine, 3-brom-2-methylanilin
PubChem CID 123538
IUPAC Name 3-bromo-2-methylaniline
SMILES CC1=C(C=CC=C1Br)N

Specifications

Color Brown
Boiling Point 255°C
Quantity 5 g
Formula Weight 186.05
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Bromo-2-methylaniline