missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-methylaniline, 98+%

Catalog No. AAA1188422
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1188422 100 g
AAA1188406 5 g
AAA1188414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1188422 Supplier Thermo Scientific Chemicals Supplier No. A1188422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 55289-36-6 | C7H8BrN | 186.052 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55289-36-6
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.052
MDL Number MFCD00051579
InChI Key IILVSKMKMOJHMA-UHFFFAOYSA-N
Synonym 2-amino-6-bromotoluene, 3-bromo-o-toluidine, benzenamine, 3-bromo-2-methyl, 3-bromo-2-methylbenzenamine, 2-bromo-6-aminotoluene, 3-bromo-2-methyl-phenylamine, 3-bromo-2-methyl-aniline, 3-bromo-2-methylphenylamine, 3-bromo-2-methyl-benzenamine, 3-brom-2-methylanilin
PubChem CID 123538
IUPAC Name 3-bromo-2-methylaniline
SMILES CC1=C(C=CC=C1Br)N

Specifications

Density 1.51
Boiling Point 253°C to 255°C
Flash Point >110°C (230°F)
Refractive Index 1.616
Quantity 100 g
UN Number UN2810
Beilstein 2354213
Sensitivity Light sensitive
Formula Weight 186.06
Percent Purity ≥98%
Chemical Name or Material 3-Bromo-2-methylaniline
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 259-568-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only