missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-methyl-1-propene 95.0+%, TCI America™

Catalog No. B24675G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B24675G Supplier TCI America Supplier No. B24675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1458-98-6
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135.00
MDL Number MFCD00134155
InChI Key USEGQJLHQSTGHW-UHFFFAOYSA-N
Synonym 3-bromo-2-methylpropene, 3-bromo-2-methyl-1-propene, methallyl bromide, 2-bromomethyl prop-1-ene, 2-methylallyl bromide, 1-propene, 3-bromo-2-methyl, 3-bromo-2-methyl-prop-1-ene, 3-bromo-2-methylpropene, stabilized with hydroquinone, methallylbromide, methylallyl bromide
PubChem CID 357785
IUPAC Name 3-bromo-2-methylprop-1-ene
SMILES CC(=C)CBr

Specifications

Color Yellow
Boiling Point 95°C
Quantity 5 g
UN Number 2924
Formula Weight 135.00
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-2-methyl-1-propene
TSCA No
Recommended Storage Refrigerator