missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-methoxyaniline 98.0+%, TCI America™

Catalog No. p-7123975
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51311G 1 g
B51315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51311G Supplier TCI America Supplier No. B51311G

Chemical Identifiers

CAS 116557-46-1
Molecular Formula C7H8BrNO
Molecular Weight (g/mol) 202.051
MDL Number MFCD13186737
InChI Key ZLODWCIXZJMLJL-UHFFFAOYSA-N
Synonym 2-Amino-6-bromoanisole, 3-Bromo-o-anisidine
PubChem CID 22667735
IUPAC Name 3-bromo-2-methoxyaniline
SMILES COC1=C(C=CC=C1Br)N

Specifications

Boiling Point 159°C
Quantity 1 g
UN Number 2810
Formula Weight 202.05
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Bromo-2-methoxyaniline