missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-hydroxy-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 76041-73-1
Molecular Formula C6H3BrF3NO
Molecular Weight (g/mol) 241.99
MDL Number MFCD02691223
InChI Key UWHKLLYQUQPOQK-UHFFFAOYSA-N
Synonym 3-bromo-5-trifluoromethyl pyridin-2-ol, 3-bromo-2-hydroxy-5-trifluoromethyl pyridine, 3-bromo-2-hydroxy-5-trifluoromethylpyridine, 3-bromo-5-trifluoromethyl pyridin-2 1h-one, 3-bromo-5-trifluoromethyl pyridine-2-ol, 3-bromo-5-trifluoromethyl-2-pyridinol, abbypharma ap-12-5330, 2 1h-pyridinone, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluoromethyl-1h-pyridin-2-one, pubchem14531
PubChem CID 13491775
IUPAC Name 3-bromo-5-(trifluoromethyl)-1H-pyridin-2-one
SMILES C1=C(C(=O)NC=C1C(F)(F)F)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC432560050
View Documents Promotion Details
N/A 5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

76041-73-1
241.99
UWHKLLYQUQPOQK-UHFFFAOYSA-N
13491775
C1=C(C(=O)NC=C1C(F)(F)F)Br
C6H3BrF3NO
MFCD02691223
3-bromo-5-trifluoromethyl pyridin-2-ol, 3-bromo-2-hydroxy-5-trifluoromethyl pyridine, 3-bromo-2-hydroxy-5-trifluoromethylpyridine, 3-bromo-5-trifluoromethyl pyridin-2 1h-one, 3-bromo-5-trifluoromethyl pyridine-2-ol, 3-bromo-5-trifluoromethyl-2-pyridinol, abbypharma ap-12-5330, 2 1h-pyridinone, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluoromethyl-1h-pyridin-2-one, pubchem14531
3-bromo-5-(trifluoromethyl)-1H-pyridin-2-one

Specifications

76041-73-1
100.0
96% min. (HPLC)
MFCD02691223
UWHKLLYQUQPOQK-UHFFFAOYSA-N
3-bromo-5-(trifluoromethyl)-1H-pyridin-2-one
13491775
97%
96.0
Authentic
C6H3BrF3NO
3-bromo-5-trifluoromethyl pyridin-2-ol, 3-bromo-2-hydroxy-5-trifluoromethyl pyridine, 3-bromo-2-hydroxy-5-trifluoromethylpyridine, 3-bromo-5-trifluoromethyl pyridin-2 1h-one, 3-bromo-5-trifluoromethyl pyridine-2-ol, 3-bromo-5-trifluoromethyl-2-pyridinol, abbypharma ap-12-5330, 2 1h-pyridinone, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluoromethyl-1h-pyridin-2-one, pubchem14531
C1=C(C(=O)NC=C1C(F)(F)F)Br
241.99
241.99
3-Bromo-2-hydroxy-5-(trifluoromethyl)pyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only