missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-2-hydroxy-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 432560050
| Quantity | 5g |
|---|---|
| Packaging | Glass bottle |
Chemical Identifiers
| 76041-73-1 | |
| 241.99 | |
| UWHKLLYQUQPOQK-UHFFFAOYSA-N | |
| 13491775 | |
| C1=C(C(=O)NC=C1C(F)(F)F)Br |
| C6H3BrF3NO | |
| MFCD02691223 | |
| 3-bromo-5-trifluoromethyl pyridin-2-ol, 3-bromo-2-hydroxy-5-trifluoromethyl pyridine, 3-bromo-2-hydroxy-5-trifluoromethylpyridine, 3-bromo-5-trifluoromethyl pyridin-2 1h-one, 3-bromo-5-trifluoromethyl pyridine-2-ol, 3-bromo-5-trifluoromethyl-2-pyridinol, abbypharma ap-12-5330, 2 1h-pyridinone, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluoromethyl-1h-pyridin-2-one, pubchem14531 | |
| 3-bromo-5-(trifluoromethyl)-1H-pyridin-2-one |
Specifications
| 3-Bromo-2-hydroxy-5-(trifluoromethyl)pyridine | |
| 96.0 | |
| Authentic | |
| Glass bottle | |
| 5g | |
| 3-bromo-5-trifluoromethyl pyridin-2-ol, 3-bromo-2-hydroxy-5-trifluoromethyl pyridine, 3-bromo-2-hydroxy-5-trifluoromethylpyridine, 3-bromo-5-trifluoromethyl pyridin-2 1h-one, 3-bromo-5-trifluoromethyl pyridine-2-ol, 3-bromo-5-trifluoromethyl-2-pyridinol, abbypharma ap-12-5330, 2 1h-pyridinone, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluoromethyl-1h-pyridin-2-one, pubchem14531 | |
| C1=C(C(=O)NC=C1C(F)(F)F)Br | |
| 241.99 | |
| 241.99 |
| 76041-73-1 | |
| 100.0 | |
| 96% min. (HPLC) | |
| C6H3BrF3NO | |
| MFCD02691223 | |
| UWHKLLYQUQPOQK-UHFFFAOYSA-N | |
| 3-bromo-5-(trifluoromethyl)-1H-pyridin-2-one | |
| 13491775 | |
| 97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash
GHS Signal Word: Warning