missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-2-fluoropyridine 98.0+%, TCI America™
$65.46 - $190.59
Chemical Identifiers
| CAS | 36178-05-9 |
|---|---|
| Molecular Formula | C5H3BrFN |
| Molecular Weight (g/mol) | 175.988 |
| MDL Number | MFCD04112496 |
| InChI Key | KESUTBOSNOHAMK-UHFFFAOYSA-N |
| PubChem CID | 2762778 |
| IUPAC Name | 3-bromo-2-fluoropyridine |
| SMILES | C1=CC(=C(N=C1)F)Br |
Chemical Identifiers
| 36178-05-9 | |
| 175.988 | |
| KESUTBOSNOHAMK-UHFFFAOYSA-N | |
| 3-bromo-2-fluoropyridine |
| C5H3BrFN | |
| MFCD04112496 | |
| 2762778 | |
| C1=CC(=C(N=C1)F)Br |
Specifications
| 36178-05-9 | |
| 76°C | |
| MFCD04112496 | |
| 2-fluoro-3-bromopyridine, 3-bromo-2-fluoro-pyridine, pyridine, 3-bromo-2-fluoro, bromofluoropyridine, pubchem3008, acmc-1ct2w, pyridine,3-bromo-2-fluoro, ksc576a4j, 3-bromo-2-fluoro pyridine | |
| C1=CC(=C(N=C1)F)Br | |
| 175.988 | |
| 175.99 | |
| Liquid |
| Yellow | |
| C5H3BrFN | |
| 1 g | |
| KESUTBOSNOHAMK-UHFFFAOYSA-N | |
| 3-bromo-2-fluoropyridine | |
| 2762778 | |
| ≥98.0% (GC) | |
| 3-Bromo-2-fluoropyridine |
Safety and Handling
TSCA : No