accessibility menu, dialog, popup

UserName

CAS RN 36178-05-9

FNBr

CAS RN 36178-05-9

IUPAC Name: 3-bromo-2-fluoropyridine
Synonyms: 3-bromo-2-fluoropyridine
Molecular Weight (g/mol): 175.99
Molecular Formula: C5H3BrFN
InChi Key: KESUTBOSNOHAMK-UHFFFAOYSA-N
SMILES: FC1=NC=CC=C1Br
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results