missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-fluorobenzyl bromide, 97%, Thermo Scientific Chemicals

$473.10 - $473.10

Chemical Identifiers

CAS 149947-16-0
Molecular Formula C7H5Br2F
Molecular Weight (g/mol) 267.923
MDL Number MFCD09842442
InChI Key MGVHFTWDCYIBDO-UHFFFAOYSA-N
PubChem CID 18989440
IUPAC Name 1-bromo-3-(bromomethyl)-2-fluorobenzene
SMILES C1=CC(=C(C(=C1)Br)F)CBr
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3222806
View Documents
Thermo Scientific Chemicals
H3222806
5 g
Each for $473.10
Only null left
 
Description

Description

Used to obtain the target inhibitors. A new quasi-one-dimensional molecular solid based on Ni (mnt) 2 anion: Crystal structure and spin-gap transition 4-Bromo-2-fluorobenzyl bromide acts as stasrting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to obtain the target inhibitors. A new quasi-one-dimensional molecular solid based on Ni (mnt) 2 anion: Crystal structure and spin-gap transition 4-Bromo-2-fluorobenzyl bromide acts as stasrting material.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

149947-16-0
267.923
MGVHFTWDCYIBDO-UHFFFAOYSA-N
1-bromo-3-(bromomethyl)-2-fluorobenzene
C7H5Br2F
MFCD09842442
18989440
C1=CC(=C(C(=C1)Br)F)CBr

Specifications

149947-16-0
C7H5Br2F
5 g
1-bromo-3-bromomethyl-2-fluorobenzene, 3-bromo-2-fluorobenzyl bromide, benzene,1-bromo-3-bromomethyl-2-fluoro, acmc-20ahso, intermediates-zcf02146, benzene, 1-bromo-3-bromomethyl-2-fluoro
C1=CC(=C(C(=C1)Br)F)CBr
267.923
267.92
3-Bromo-2-fluorobenzyl bromide
60°C (0.1 mmHg)
MFCD09842442
UN3265
MGVHFTWDCYIBDO-UHFFFAOYSA-N
1-bromo-3-(bromomethyl)-2-fluorobenzene
18989440
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only