missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-fluorobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog No. AAH3222806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3222806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3222806 Supplier Thermo Scientific Chemicals Supplier No. H3222806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Used to obtain the target inhibitors. A new quasi-one-dimensional molecular solid based on Ni (mnt) 2 anion: Crystal structure and spin-gap transition 4-Bromo-2-fluorobenzyl bromide acts as stasrting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to obtain the target inhibitors. A new quasi-one-dimensional molecular solid based on Ni (mnt) 2 anion: Crystal structure and spin-gap transition 4-Bromo-2-fluorobenzyl bromide acts as stasrting material.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 149947-16-0
Molecular Formula C7H5Br2F
Molecular Weight (g/mol) 267.923
MDL Number MFCD09842442
InChI Key MGVHFTWDCYIBDO-UHFFFAOYSA-N
PubChem CID 18989440
IUPAC Name 1-bromo-3-(bromomethyl)-2-fluorobenzene
SMILES C1=CC(=C(C(=C1)Br)F)CBr

Specifications

Boiling Point 60°C (0.1 mmHg)
Quantity 5 g
UN Number UN3265
Synonym 1-bromo-3-bromomethyl-2-fluorobenzene, 3-bromo-2-fluorobenzyl bromide, benzene,1-bromo-3-bromomethyl-2-fluoro, acmc-20ahso, intermediates-zcf02146, benzene, 1-bromo-3-bromomethyl-2-fluoro
Formula Weight 267.92
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-fluorobenzyl bromide
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only