missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-fluorobenzoic Acid 98.0+%, TCI America™

Catalog No. B39295G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39295G 5 g
B39291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39295G Supplier TCI America Supplier No. B39295G

Chemical Identifiers

CAS 161957-56-8
Molecular Formula C7H4BrFO2
Molecular Weight (g/mol) 219.009
MDL Number MFCD00665763
InChI Key UVKURTLVTLRSSM-UHFFFAOYSA-N
Synonym 3-bromo-2-fluoro-benzoic acid, 3-bromo-2-fluorobenzoicacid, 2-fluoro-3-bromobenzoic acid, benzoic acid, 3-bromo-2-fluoro, 3-bromo-2-fluoro benzoic acid, buttpark 14\01-30, 3-bromo-2-fluorobenzenecarboxylic acid, pubchem1318, acmc-209dnk, intermediates-zcf02220
PubChem CID 2736313
IUPAC Name 3-bromo-2-fluorobenzoic acid
SMILES C1=CC(=C(C(=C1)Br)F)C(=O)O

Specifications

Melting Point 170°C
Color White-Yellow
Quantity 5 g
Formula Weight 219.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-2-fluorobenzoic Acid